Learn More
Perkin Elmer US LLC 3-Slot Demountable Quartz Torch for Optima 2x00/4x00/5x00/7x00 DV
Catalog No. 509047597
Encompass_Preferred
This demountable 3-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow and is recommended for use in organic analysis.
Catalog No. 50-904-7597 Supplier Perkin Elmer US LLC Supplier No. N0772005
This demountable 3-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow and is recommended for use in organic analysis.