Learn More
Glass Expansion Torch Adaptor for Injector without Ball Joint. Cassette Mount Adaptor for Injector without ball joint. This adaptor is suitable for injectors without a ball joint. Compatible with PerkinElmer Elan non-DRC versions, NexION 300/350.
SDP
Catalog No. 502703153
Encompass_Preferred
PFA Cassette Mount Adaptor 15mm (W1013266). This adaptor is suitable for injectors without a ball joint. This torch adaptor is usually used with non-DRC PerkinElmer Elan models, or with Elan DRC models using an HF-resistant sample introduction system. It requires the Elan Screw Mount CSC Adaptor P/N 21-808-0389.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3153 Supplier Glass Expansion Supplier No. 318082825
PFA Cassette Mount Adaptor 15mm (W1013266). This adaptor is suitable for injectors without a ball joint. This torch adaptor is usually used with non-DRC PerkinElmer Elan models, or with Elan DRC models using an HF-resistant sample introduction system. It requires the Elan Screw Mount CSC Adaptor P/N 21-808-0389.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001