Learn More
Perkin Elmer US LLC Torch Cassette, Platinum - Cassette Only
Catalog No. 502111618
Encompass_Preferred
This cassette is included in the NexION 2000 ICP-MS PFA-Platinum HF-Resistant Sample Introduction Kit (N8150033) and the Platinum SMARTintro Sample Introduction Module (N8152441).
Catalog No. 50-211-1618 Supplier Perkin Elmer US LLC Supplier No. N8152434
This cassette is included in the NexION 2000 ICP-MS PFA-Platinum HF-Resistant Sample Introduction Kit (N8150033) and the Platinum SMARTintro Sample Introduction Module (N8152441).