Learn More
Agilent Technologies Torch Cleaning Stand, 5000 Series ICP-OES, 1/Pk.
Catalog No. NC2942514
Encompass
This torch cleaning stand provides a stable platform designed to facilitate the maintenance of ICP-OES torches. It allows users to safely suspend either the complete torch or the individual injector and base assembly within a cleaning solution.
  • Supports safe and stable suspension of torch components during cleaning
  • Reduces the risk of spills and accidental torch damage
  • Compatible with 5000 series ICP-OES torch systems
  • Suitable for 5100, 5110, 5800, and 5900 instrument models
Catalog No. NC2942514 Supplier Agilent Technologies Supplier No. G801068021
This torch cleaning stand provides a stable platform designed to facilitate the maintenance of ICP-OES torches. It allows users to safely suspend either the complete torch or the individual injector and base assembly within a cleaning solution.
  • Supports safe and stable suspension of torch components during cleaning
  • Reduces the risk of spills and accidental torch damage
  • Compatible with 5000 series ICP-OES torch systems
  • Suitable for 5100, 5110, 5800, and 5900 instrument models