Learn More
Perkin Elmer US LLC 0-Slot High Efficiency Demountable Quartz Torch for ELAN 9000/6x00/DRCs
Catalog No. 509052392
Encompass_Preferred
Demountable design using one-piece quartz tubing for plasma and auxiliary gas flow. The torch operates at 25% reduced argon gas flows compared to the standard demountable quartz torch (N8122006). The High Efficiency Quartz Torch Installation Kit (W1007468) is required for first-time installations of the high efficiently quartz torch for ELAN 9000 and 6x00 series instruments.
Catalog No. 50-905-2392 Supplier Perkin Elmer US LLC Supplier No. W1008384
Demountable design using one-piece quartz tubing for plasma and auxiliary gas flow. The torch operates at 25% reduced argon gas flows compared to the standard demountable quartz torch (N8122006). The High Efficiency Quartz Torch Installation Kit (W1007468) is required for first-time installations of the high efficiently quartz torch for ELAN 9000 and 6x00 series instruments.