Learn More
Thermo Scientific TORCH HOLDER ASSEMBLY SPARES
Supplier:  Thermo Scientific 842315550171
Encompass
Torch Holder (EMT Torch); Accommodates the EMT torch and EMT centre tube holder and provides a; secure mount to the hold the EMT torch in the iCAP torch box.
Catalog No. NC9819376