Learn More
Perkin Elmer US LLC 0-Slot Demountable Quartz Short Torch for Optima 2x00/4x00/5x00/7x00 DV
Catalog No. 509043781
Encompass_Preferred
This demountable 0-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow and is recommended for use with organic solvents. It is the short version of the regular 0-Slot Demountable Quartz Torch (N0770343).
Catalog No. 50-904-3781 Supplier Perkin Elmer US LLC Supplier No. N0770344
This demountable 0-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow and is recommended for use with organic solvents. It is the short version of the regular 0-Slot Demountable Quartz Torch (N0770343).