Learn More
Selleck Chemical LLC Toremifene Citrate (NK 622) S1776-10mM/1mL
SDP
Catalog No. 503110603
Encompass_Preferred
Toremifene Citrate (NK 622 NSC 613680) is an oral selective estrogen receptor modulator (SERM) used in the treatment of advanced breast cancer
Catalog No. 50-311-0603 Supplier Selleck Chemical LLC Supplier No. S177610MM/1ML
Toremifene Citrate (NK 622 NSC 613680) is an oral selective estrogen receptor modulator (SERM) used in the treatment of advanced breast cancer