Learn More
Apexbio Technology LLC Toremifene Citrate 89778-27-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000088211
Encompass_Preferred
Toremifene Citrate (CAS 89778-27-8) is a small-molecule selective estrogen receptor modulator (SERM) targeting estrogen receptors It is designed to modulate estrogen receptor signaling pathways thereby regulating estrogen-dependent cellular functions Toremifene Citrate exerts its biological activity primarily through tissue-specific antagonism or partial agonism of the estrogen receptor In in vitro studies Toremifene Citrate demonstrates receptor binding activity with an affinity of 1 M for the estrogen receptor Based on these pharmacological properties Toremifene Citrate holds research potential in investigating estrogen-dependent regulatory processes and endocrine signaling dynamics
Catalog No. 50-000-88211 Supplier Apexbio Technology LLC Supplier No. B15135.1
Toremifene Citrate (CAS 89778-27-8) is a small-molecule selective estrogen receptor modulator (SERM) targeting estrogen receptors It is designed to modulate estrogen receptor signaling pathways thereby regulating estrogen-dependent cellular functions Toremifene Citrate exerts its biological activity primarily through tissue-specific antagonism or partial agonism of the estrogen receptor In in vitro studies Toremifene Citrate demonstrates receptor binding activity with an affinity of 1 M for the estrogen receptor Based on these pharmacological properties Toremifene Citrate holds research potential in investigating estrogen-dependent regulatory processes and endocrine signaling dynamics