Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toremifene Citrate 98.0+%, TCI America™
SDP

Catalog No. T28321G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28321G 1 g
T28325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T28321G Supplier TCI America Supplier No. T28321G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 89778-27-8
Molecular Formula C32H36ClNO8
Molecular Weight (g/mol) 598.089
MDL Number MFCD01729016
InChI Key IWEQQRMGNVVKQW-OQKDUQJOSA-N
Synonym (Z)-4-Chloro-1-(4-dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene Citrate, (Z)-2-[4-(4-Chloro-1,2-diphenyl-1-butenyl)phenoxy]-N,N-dimethylethanamine Citrate
PubChem CID 3005572
IUPAC Name 2-[4-[(Z)-4-chloro-1,2-diphenylbut-1-enyl]phenoxy]-N,N-dimethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES CN(C)CCOC1=CC=C(C=C1)C(=C(CCCl)C2=CC=CC=C2)C3=CC=CC=C3.C(C(=O)O)C(CC(=O)O)(C(=O)O)O

Specifications

Melting Point 162°C
Color White-Yellow
Quantity 1 g
Formula Weight 598.09
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Toremifene Citrate
RTECSNumber KH2156700
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.