Learn More
Medchemexpress LLC HY-13003 50mg , Torin 1 CAS:1222998-36-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1300350MG
Encompass_Preferred
HY-13003 50mg Torin 1 CAS: 1222998-36-8 Torin 1 is a potent inhibitor of mTOR with an IC50 of 3 nM. Torin 1 inhibits both mTORC1/2 complexes with IC50 values between 2 and 10 nM. Torin 1 is an effective inducer of autophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0027