Learn More
Ubpbio Torin-1, 5MG
Supplier:  Ubpbio F6100
Encompass_Preferred
Torin 1-is a cell permeable highly potent, selective and ATP-competitive mTOR inhibitor of the mammalian target of rapamycin (mTOR) kinase. Torin-1 exhibits over 100-fold selectivity for mTOR over PI3Kα and other PI3K-like kinases, as well as 450 other protein kinases. Torin-1 has been found to impair cell growth and proliferation through a mechanism involving mTORC1 inhibition other than mTORC2 inhibition, in which the rapamycin-resistant functions of mTORC1 is suppressed.
Catalog No. 50-196-5118