Learn More
Apexbio Technology LLC Torin 1, 1222998-36-8, MFCD18782653, 5 mg
SDP
Supplier:  Apexbio Technology LLC A83125
Encompass_Preferred
MF: C35H28F3N5O2, MW: 607.64, Solubility: insoluble in DMSO, 2.42 mg/mL in EtOH with ultrasonic and warming,insoluble in H2O. Torin 1, a tricyclic benzonaphthyridinone, is a potent and selective inhibitor of the mammalian target of rapamycin (mTOR) kinase, which is the catalytic subunit of two functionally distinct complexes and plays a pivotal role in the regulation of cell growth, proliferation and survival.
Catalog No. 50-101-4432