Learn More
Selleck Chemical LLC Torin 2 5mg 1223001-51-1
SDP
Supplier:  Selleck Chemical LLC S28175MG
Encompass_Preferred
Torin 2 is a potent and selective mTOR inhibitor with IC50 of 0.25 nM in p53?/? MEFs cell line; 800-fold greater selectivity for mTOR than PI3K and improved pharmacokinetic properties. Inhibition of ATM/ATR/DNA-PK with EC50 of 28 nM/35 nM/118 nM,in PC3 cell lines respectively. Torin 2 decreases cell viability and induces autophagy and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3009