Learn More
Medchemexpress LLC Toringin | 1329-10-8 | 99.6% | 416.38 g/mol | C21H20O9 | 10 MG
SDP
Catalog No. 5000289481
Encompass_Preferred
Toringin is a natural bioflavonoid isolated from the bark of Docyniopsis tschonoski, supplied for research use as a white-to-yellow solid (CAS 1329-10-8). The compound is offered in small research pack sizes and is accompanied by a product data sheet and safety data sheet.
  • Natural bioflavonoid isolated from Docyniopsis tschonoski.
  • White to yellow solid suitable for analytical and biological research.
  • Reported high purity (approximately 99.6%).
  • Supplied as a 10 mg vial for small-scale experiments.
  • Documentation available: product data sheet and safety data sheet.
  • Recommended storage: powder at -20°C; in solvent, -80°C for long-term storage.
Catalog No. 50-002-89481 Supplier Medchemexpress LLC Supplier No. HYN419210MG
Toringin is a natural bioflavonoid isolated from the bark of Docyniopsis tschonoski, supplied for research use as a white-to-yellow solid (CAS 1329-10-8). The compound is offered in small research pack sizes and is accompanied by a product data sheet and safety data sheet.
  • Natural bioflavonoid isolated from Docyniopsis tschonoski.
  • White to yellow solid suitable for analytical and biological research.
  • Reported high purity (approximately 99.6%).
  • Supplied as a 10 mg vial for small-scale experiments.
  • Documentation available: product data sheet and safety data sheet.
  • Recommended storage: powder at -20°C; in solvent, -80°C for long-term storage.