Learn More
Enterprise Technology Solutions TORK Cloths,f/Cleaning,Industrial,4-1/5"x14",120/PK,4PK/CT,GY
SDP
Catalog No. 5000829361
Encompass_Preferred
Industrial Cleaning Cloths are soft and flexible enough to clean tight spaces or intricate components without scratching surfaces. They are tough enough to wipe up most oil, grease, dirt and most solvents. Use them for light cleaning or medium cleaning tasks. ExcelCLEAN technology reduces solvent consumption by up to 40 percent and helps minimize your environmental footprint. Single dispensing reduces towel waste. FSC-certified cloths are compatible with Tork Folded Wiper/Cloth Dispensers (none included). They measure 14" x 16-3/10" when unfolded.
Catalog No. 50-008-29361 Supplier Enterprise Technology Solutions Supplier No. TRK520681
Industrial Cleaning Cloths are soft and flexible enough to clean tight spaces or intricate components without scratching surfaces. They are tough enough to wipe up most oil, grease, dirt and most solvents. Use them for light cleaning or medium cleaning tasks. ExcelCLEAN technology reduces solvent consumption by up to 40 percent and helps minimize your environmental footprint. Single dispensing reduces towel waste. FSC-certified cloths are compatible with Tork Folded Wiper/Cloth Dispensers (none included). They measure 14" x 16-3/10" when unfolded.