Learn More
Enterprise Technology Solutions TORK Hand Towel Roll, 7-3/4"x700", 6/CT, White
SDP
Catalog No. 5000828821
Encompass_Preferred
Designed specifically for the Tork Elevation Matic hand towel roll dispenser (sold separately), these highly absorbent towels offer the perfect combination of quality, performance and value. Embossing enhances hand feel and maximum absorbency so fewer towels are used per hand-dry. Less packaging reduces your overall labor and maintenance costs. Long-lasting rolls minimize maintenance costs while maximizing service. Eco-friendly design is made from a high percentage of post-consumer fiber to provide environmental benefits and help you create a positive image.
Catalog No. 50-008-28821 Supplier Enterprise Technology Solutions Supplier No. TRK290089
Designed specifically for the Tork Elevation Matic hand towel roll dispenser (sold separately), these highly absorbent towels offer the perfect combination of quality, performance and value. Embossing enhances hand feel and maximum absorbency so fewer towels are used per hand-dry. Less packaging reduces your overall labor and maintenance costs. Long-lasting rolls minimize maintenance costs while maximizing service. Eco-friendly design is made from a high percentage of post-consumer fiber to provide environmental benefits and help you create a positive image.