Learn More
Medchemexpress LLC HY-10474 10mg Medchemexpress, Torkinib CAS:1092351-67-1 Purity:>98%
SDP
Catalog No. 502028759
Encompass_Preferred
Medchemexpress, HY-10474 10mg Torkinib CAS:1092351-67-1 Torkinib (PP 242) is a selective and ATP-competitive mTOR inhibitor with an IC50 of 8 nM. PP242 inhibits both mTORC1 and mTORC2 with IC50s of 30 nM and 58 nM, respectively[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8759 Supplier Medchemexpress LLC Supplier No. HY1047410MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-10474 10mg Torkinib CAS:1092351-67-1 Torkinib (PP 242) is a selective and ATP-competitive mTOR inhibitor with an IC50 of 8 nM. PP242 inhibits both mTORC1 and mTORC2 with IC50s of 30 nM and 58 nM, respectively[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.