Learn More
TARGETMOL CHEMICALS INC TORKINIB 50MG
SDP
Catalog No. 502577127
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Torkinib (PP 242) (PP 242) is a selective and ATP-competitive mTOR inhibitor (IC50 8 nM). It also inhibits mTORC1/2 (IC50s 30/58 nM). purity: 99%
Catalog No. 50-257-7127 Supplier TARGETMOL CHEMICALS INC Supplier No. T241450MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Torkinib (PP 242) (PP 242) is a selective and ATP-competitive mTOR inhibitor (IC50 8 nM). It also inhibits mTORC1/2 (IC50s 30/58 nM). purity: 99%