Learn More
KINEX CAPPERS LLC Tornado Digital Torque Tester; 0 - 13 inch pounds
Supplier: KINEX CAPPERS LLC MT876103
Key FeaturesTorque range: 0 - 13 inch pounds Accuracy: +/- 0.5% of full scaleBottle Diameter: 0.4 - 7.5 inches (10-190mm)Clockwise & counter-clockwise digital torque captureEasy export to a PC, printer, etc, via RS232 portRugged, water resistant case (rated to IP 54)Includes a Certificate of CalibrationHighly Portable: molded carry handle and compact, lightweight designPlug into an outlet or use the on board batteryPlusTamper Evident Closure TestingOn board memory: store up to 500 resultsAudible and Visual Pass/Fail Alarms