Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tornado Tube
SCI_ED

Catalog No. S41568
Change view
Click to view available options
Product Type:
Tornado Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S41568 Tornado Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S41568 Supplier Tornado Tube Supplier No. #200MN96

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Simulate hydraulic conditions that create tornadoes.

  • Coupler attaches two 16 oz., 1L or 2L screw-top soda bottles together
  • Rotating water in the top bottle simulates conditions that create tornadoes
  • As water falls from the top to the bottom bottle, water moves in a tornado-like shape
  • Brief explanation of hydraulic principles is printed on the package

Specifications

Weight (Metric) 0.45 kg
Product Type Tornado Tube
Weight (English) 0.99 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.