Learn More
Grainger Toroidal Conductivity Kit Noryl
Catalog No. 502768020
Encompass_Preferred
The SensoRex Toroidal Conductivity Kit is a portable conductivity meter kit designed for water quality testing. It includes a toroidal conductivity sensor, calibration solutions, carrying case, and 9V battery. The meter has a measurement range of 0 to 1999 μS/cm, resolution of 1 μS/cm, and accuracy of ±2% full scale. The toroidal sensor provides flow-through measurement and resists polarization and fouling. This kit is ideal for testing drinking water, wastewater, process water, and more. The rugged carrying case allows for convenient field use.
Catalog No. 50-276-8020 Supplier Grainger Supplier No. 16Z069
The SensoRex Toroidal Conductivity Kit is a portable conductivity meter kit designed for water quality testing. It includes a toroidal conductivity sensor, calibration solutions, carrying case, and 9V battery. The meter has a measurement range of 0 to 1999 μS/cm, resolution of 1 μS/cm, and accuracy of ±2% full scale. The toroidal sensor provides flow-through measurement and resists polarization and fouling. This kit is ideal for testing drinking water, wastewater, process water, and more. The rugged carrying case allows for convenient field use.