Learn More
Grainger Toroidal Conductivity Kit Noryl
Catalog No. 502768178
Encompass_Preferred
The SensoRex Toroidal Conductivity Kit is a portable conductivity meter kit designed for water quality testing. It includes a toroidal conductivity sensor, calibration solutions, plastic beakers, plastic pipettes, and a hard carrying case. The meter has a measurement range of 0 to 1999 μS/cm. Key features include one-button calibration, auto-ranging, auto-off, and replaceable sensors. This kit contains everything needed for quick and accurate on-site water conductivity measurements. With a compact design and rugged case, it is well-suited for field applications.
Catalog No. 50-276-8178 Supplier Grainger Supplier No. 16Z068
The SensoRex Toroidal Conductivity Kit is a portable conductivity meter kit designed for water quality testing. It includes a toroidal conductivity sensor, calibration solutions, plastic beakers, plastic pipettes, and a hard carrying case. The meter has a measurement range of 0 to 1999 μS/cm. Key features include one-button calibration, auto-ranging, auto-off, and replaceable sensors. This kit contains everything needed for quick and accurate on-site water conductivity measurements. With a compact design and rugged case, it is well-suited for field applications.