Learn More
Grainger Toroidal Conductivity Sensor Noryl
Catalog No. 502768167
Encompass_Preferred
The SENSOREX TCS3200 Toroidal Conductivity Sensor is an industrial conductivity sensor designed for continuous monitoring and control of conductivity in pure and ultrapure water applications. It features a toroidal measuring cell made of PEEK with platinum electrodes for corrosion resistance. The large flow-through design allows for representative sampling across the entire pipe diameter. Measurement range is 0.055 to 1000 μS/cm with accuracy of ±1% of reading. The sensor provides analog 4-20mA and digital MODBUS RTU output. Maximum pressure is 145 psi at 104°F. Applications include semiconductor manufacturing, power generation, pharmaceutical production, and laboratory use.
Catalog No. 50-276-8167 Supplier Grainger Supplier No. 16Z065
The SENSOREX TCS3200 Toroidal Conductivity Sensor is an industrial conductivity sensor designed for continuous monitoring and control of conductivity in pure and ultrapure water applications. It features a toroidal measuring cell made of PEEK with platinum electrodes for corrosion resistance. The large flow-through design allows for representative sampling across the entire pipe diameter. Measurement range is 0.055 to 1000 μS/cm with accuracy of ±1% of reading. The sensor provides analog 4-20mA and digital MODBUS RTU output. Maximum pressure is 145 psi at 104°F. Applications include semiconductor manufacturing, power generation, pharmaceutical production, and laboratory use.