Learn More
MSC Klein Tools Torpedo Level, Billet Aluminum, 8 in., Magnetic, 1 ea.
Catalog No. 5000514880
Encompass
This 8 in. torpedo level is a durable, professional-grade measuring tool constructed from billet aluminum. Designed for high-visibility and ease of use in diverse field conditions, it features a specialized magnetic track and multiple high-visibility vials for precise leveling applications.
  • Constructed from billet aluminum for enhanced durability
  • Features four high-visibility vials for 0°, 90°, 45°, and 30° measurements
  • Equipped with Rare Earth magnets in a patented track to prevent magnet loss
  • High-visibility orange body for easy identification on jobsites
  • V-groove design fits the contour of conduit and pipe
  • Tapered nose allows for convenient pocket storage
  • Accuracy of ±0.029°
Catalog No. 50-005-14880 Supplier MSC Supplier No. 49286834
This 8 in. torpedo level is a durable, professional-grade measuring tool constructed from billet aluminum. Designed for high-visibility and ease of use in diverse field conditions, it features a specialized magnetic track and multiple high-visibility vials for precise leveling applications.
  • Constructed from billet aluminum for enhanced durability
  • Features four high-visibility vials for 0°, 90°, 45°, and 30° measurements
  • Equipped with Rare Earth magnets in a patented track to prevent magnet loss
  • High-visibility orange body for easy identification on jobsites
  • V-groove design fits the contour of conduit and pipe
  • Tapered nose allows for convenient pocket storage
  • Accuracy of ±0.029°