Learn More
Grainger Torque Driver Set 1/4 10 to 50 -lb.
Catalog No. 502768585
Encompass_Preferred
The SK Professional Tools Torque Screwdriver Set is a set of 6 precision screwdrivers with torque limiting handles that allow the user to accurately tighten screws to a preset torque level. The set includes 6 screwdrivers with torque settings from 2.5 to 10 inch-pounds. The screwdrivers have chrome vanadium steel blades and soft grip handles. This set is ideal for electronics repair and other applications requiring precise torque control. The screwdrivers are made in the USA and come with a lifetime warranty from the manufacturer.
Catalog No. 50-276-8585 Supplier Grainger Supplier No. 170F99
The SK Professional Tools Torque Screwdriver Set is a set of 6 precision screwdrivers with torque limiting handles that allow the user to accurately tighten screws to a preset torque level. The set includes 6 screwdrivers with torque settings from 2.5 to 10 inch-pounds. The screwdrivers have chrome vanadium steel blades and soft grip handles. This set is ideal for electronics repair and other applications requiring precise torque control. The screwdrivers are made in the USA and come with a lifetime warranty from the manufacturer.