Learn More
Scientific Industries TORQUE GAU TORBAL FSA2 17LBF
Supplier:  Scientific Industries FSA2
Encompass_Preferred
17lbf.in x 0.01lbf.in (2Nm x 0.001Nm) FSA2 TORQUE is ideal for any clockwise and counter clockwise cap torque testing application. Easily test any cap, jar, bottle, or valve. Fully configurable and easy-to-use Peak modes increase efficiency during quality control testing or research. Ultra-fast real time 1000Hz sampling rate guarantees accurate and precise torque measurements. Every model is equipped with a built-in wireless data transmitter, expandable microSD memory card, as well as a USB and an RS232 interface. Every function is optimized with statistical analysis, onscreen graphs, and a histogram. The analog input-output port makes the FSA ideal for use with Pass / Fail signalization devices or other equipment. Engineered with high output loadcells to deliver repeatable torque measurements at all times. The FSA is built with a heavy-duty stable die-cast metal housing and designed to accept containers or samples up to 7” in diameter.
Catalog No. 50-125-2321