Learn More
MSC Proto Micrometer Torque Wrench, 3/4 in, 120-600 ft-lb, 1 ea
Catalog No. 501920509
Encompass
The Proto Micrometer Torque Wrench has a 3/4 in drive and measures torque in foot-pounds. It has a minimum torque of 120.0 ft-lb and an overall length of 41-7/16 in.
  • Wrench type: micrometer torque wrench
  • Torque measurement type: foot pound
  • Minimum torque: 120.0 ft-lb
  • Drive size: 3/4 in
  • Head type: ratchet
  • Country of origin: United States
Catalog No. 50-192-0509 Supplier MSC Supplier No. 09812272
The Proto Micrometer Torque Wrench has a 3/4 in drive and measures torque in foot-pounds. It has a minimum torque of 120.0 ft-lb and an overall length of 41-7/16 in.
  • Wrench type: micrometer torque wrench
  • Torque measurement type: foot pound
  • Minimum torque: 120.0 ft-lb
  • Drive size: 3/4 in
  • Head type: ratchet
  • Country of origin: United States