Learn More
Across International ULVAC DTC-41 110V 1.6 cfm 2-Stage Chemical-Duty Diaphragm Pump TUV
SDP
Catalog No. NC1814829
Encompass_Preferred
Manufactured and imported directly from Japan, ULVAC DTC series diaphragm pumps feature integral forced-air cooling. They have two-stage PTFE diaphragms that are ideal for pumping corrosive gases and organic solvents. These pumps are oil-free, portable, and chemical-resistant, which means less maintenance and no more frequent and costly oil change, or cross contamination.
Catalog No. NC1814829 Supplier Across International Supplier No. DTC41UL
Manufactured and imported directly from Japan, ULVAC DTC series diaphragm pumps feature integral forced-air cooling. They have two-stage PTFE diaphragms that are ideal for pumping corrosive gases and organic solvents. These pumps are oil-free, portable, and chemical-resistant, which means less maintenance and no more frequent and costly oil change, or cross contamination.