Learn More
Cole-Parmer Torrey Pines EchoTherm HS40 Programmable Digital Stirring Hot Plate, 115V
Catalog No. NC1730481 Shop All Cole-Parmer Products
Encompass_Preferred
Repeat any program up to 98 times automatically. Multiple temperatures, temperature ramp rates, stirring speeds and timed events can be stored. Includes timer to 99 hours, 59 minutes with auto-off option. Unit can be programmable or non-programmable in operation. Offers RS232 interface for control or recording data via a computer. 6-foot cord with plug is included.
Catalog No. NC1730481 Supplier Cole-Parmer Supplier No. 0464600
Repeat any program up to 98 times automatically. Multiple temperatures, temperature ramp rates, stirring speeds and timed events can be stored. Includes timer to 99 hours, 59 minutes with auto-off option. Unit can be programmable or non-programmable in operation. Offers RS232 interface for control or recording data via a computer. 6-foot cord with plug is included.