Learn More
Apexbio Technology LLC Torsemide 56211-40-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063456
Encompass_Preferred
Torsemide a pyridine-sulfonyl urea derivative acts pharmacologically as a loop diuretic by selectively inhibiting the Na -K -2Cl cotransporter (NKCC2) located in the thick ascending limb of Henle s loop Through this mechanism torsemide reduces sodium chloride and water reabsorption resulting in increased urinary excretion of electrolytes and water Chemically characterized by a sulfonylurea moiety linked to pyridine functionality torsemide is utilized in pharmacological research to investigate kidney function fluid-electrolyte balance and associated physiological responses Additionally researchers employ torsemide as a tool compound to study ion transport regulation pathways and the pathophysiology underlying edema and hypertension at a molecular and cellular level
Catalog No. 50-000-63456 Supplier Apexbio Technology LLC Supplier No. B20555.1
Torsemide a pyridine-sulfonyl urea derivative acts pharmacologically as a loop diuretic by selectively inhibiting the Na -K -2Cl cotransporter (NKCC2) located in the thick ascending limb of Henle s loop Through this mechanism torsemide reduces sodium chloride and water reabsorption resulting in increased urinary excretion of electrolytes and water Chemically characterized by a sulfonylurea moiety linked to pyridine functionality torsemide is utilized in pharmacological research to investigate kidney function fluid-electrolyte balance and associated physiological responses Additionally researchers employ torsemide as a tool compound to study ion transport regulation pathways and the pathophysiology underlying edema and hypertension at a molecular and cellular level