Learn More
Medchemexpress LLC Torsemide-d7 | 1189375-06-1 | 99.8% | 355.46 | C16H13D7N4O3S | 10 MG
SDP
Catalog No. 5000298993
Encompass_Preferred
Torsemide-d7 is the deuterium-labeled analogue of the loop diuretic torsemide, provided as a high-purity analytical reference standard for use in mass spectrometry and pharmacokinetic studies. Supplied as a 10 mg vial for use as an internal standard or tracer in LC-MS/MS workflows; for research use only.
  • Deuterium-labeled internal standard for torsemide
  • High purity 99.8% suitable for quantitative analysis
  • Molecular weight 355.46 g/mol
  • Supplied as a 10 mg vial
  • Intended for research use only, not for human or clinical use
Catalog No. 50-002-98993 Supplier Medchemexpress LLC Supplier No. HYB0247S10MG
Torsemide-d7 is the deuterium-labeled analogue of the loop diuretic torsemide, provided as a high-purity analytical reference standard for use in mass spectrometry and pharmacokinetic studies. Supplied as a 10 mg vial for use as an internal standard or tracer in LC-MS/MS workflows; for research use only.
  • Deuterium-labeled internal standard for torsemide
  • High purity 99.8% suitable for quantitative analysis
  • Molecular weight 355.46 g/mol
  • Supplied as a 10 mg vial
  • Intended for research use only, not for human or clinical use