Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tortoside A | 190655-16-4 | 1 MG
SDP

Catalog No. 5000417610
Encompass_Preferred

Tortoside A is a bioactive compound isolated from the roots of Ilex pubescens. It is available for research use, with a high purity of 98.85%. This compound is classified under phenylpropanoids and lignans, and is provided as a solid, white to off-white in appearance.

  • High purity: 98.85%
  • Naturally derived: Found in Ilex pubescens roots
  • Form: Solid, white to off-white appearance
  • Applications: For research use only
  • Storage: Powder at -20°C for 3 years; in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-17610 Supplier Medchemexpress LLC Supplier No. HYN81541MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tortoside A is a bioactive compound isolated from the roots of Ilex pubescens. It is available for research use, with a high purity of 98.85%. This compound is classified under phenylpropanoids and lignans, and is provided as a solid, white to off-white in appearance.

  • High purity: 98.85%
  • Naturally derived: Found in Ilex pubescens roots
  • Form: Solid, white to off-white appearance
  • Applications: For research use only
  • Storage: Powder at -20°C for 3 years; in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.