Learn More
Waters Corp Torus 1-AA OBD Prep Column, 130Å, 5 μm, 19 mm × 150 mm
Supplier:  Waters Corp 186008638
Encompass_Preferred
The Torus 1-AA columns are designed for separating neutral compounds like polar and non-polar steroids, and hydrophobic compounds such as lipids and fat-soluble vitamins. This chemistry also provides an orthogonal selectivity to another phase, which is useful in method development.
  • Superior choice for separating neutral compounds.
  • Effective for hydrophobic (lipophilic) compounds.
  • Useful for free fatty acids.
  • Suitable for fat-soluble vitamins.
  • Applicable for lipids.
  • Good for natural products.
  • Can be used for steroids.
  • Offers orthogonal selectivity, aiding method development.
  • Demonstrates good peak shape and resolution for fat-soluble vitamins.
Catalog No. 50-239-2704