Learn More
Waters Corp OBD Prep Column, 1-AA, 30 x 150 mm, 5 μm
Catalog No. 502392535 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Torus 1-AA OBD Prep Column is designed for efficient separation of neutral compounds, including polar and non-polar steroids, as well as hydrophobic compounds like lipids and fat-soluble vitamins. It features Ethylene Bridged Hybrid (BEH) particles with 1-Aminoanthracene functionalization.
  • 30 x 150 mm dimensions for preparative applications.
  • 5 μm particle size for optimal resolution.
  • Based on Ethylene Bridged Hybrid (BEH) particles.
  • 1-Aminoanthracene functionalization provides unique selectivity.
  • Designed for separation of neutral compounds, including polar and non-polar steroids.
  • Effective for hydrophobic compounds such as lipids and fat-soluble vitamins.
  • Offers orthogonal selectivity (S >90) to the 2-PIC phase, aiding method development.
Catalog No. 50-239-2535 Supplier Waters Corp Supplier No. 186008643
Add to Cart
Add to Cart
Request a Quote
The Torus 1-AA OBD Prep Column is designed for efficient separation of neutral compounds, including polar and non-polar steroids, as well as hydrophobic compounds like lipids and fat-soluble vitamins. It features Ethylene Bridged Hybrid (BEH) particles with 1-Aminoanthracene functionalization.
  • 30 x 150 mm dimensions for preparative applications.
  • 5 μm particle size for optimal resolution.
  • Based on Ethylene Bridged Hybrid (BEH) particles.
  • 1-Aminoanthracene functionalization provides unique selectivity.
  • Designed for separation of neutral compounds, including polar and non-polar steroids.
  • Effective for hydrophobic compounds such as lipids and fat-soluble vitamins.
  • Offers orthogonal selectivity (S >90) to the 2-PIC phase, aiding method development.