Learn More
Waters Corp VanGuard Pre-column, 1-AA, 130Å, 1.7 μm, 2.1 mm × 5 mm
Catalog No. 502390816 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Torus pre-columns are designed for use with the ACQUITY UPC2 System to achieve fast, robust achiral separations. They are intended to simplify the method development process, featuring new and innovative 1.7 μm chemistries for convergence chromatography. These pre-columns aim for excellent peak shape, reduce or eliminate the need for additives, offer added selectivity for a wide range of compounds, and provide improved robustness.
  • Specifically designed to use the complete range of capabilities for fast, robust achiral separations.
  • Simplifies the method development process with new and innovative 1.7 μm chemistries for convergence chromatography.
  • Designed for excellent peak shape that eliminates or reduces the need for additives and offers added selectivity for a wide range of compounds and improved robustness.
Catalog No. 50-239-0816 Supplier Waters Corp Supplier No. 186007631
Torus pre-columns are designed for use with the ACQUITY UPC2 System to achieve fast, robust achiral separations. They are intended to simplify the method development process, featuring new and innovative 1.7 μm chemistries for convergence chromatography. These pre-columns aim for excellent peak shape, reduce or eliminate the need for additives, offer added selectivity for a wide range of compounds, and provide improved robustness.
  • Specifically designed to use the complete range of capabilities for fast, robust achiral separations.
  • Simplifies the method development process with new and innovative 1.7 μm chemistries for convergence chromatography.
  • Designed for excellent peak shape that eliminates or reduces the need for additives and offers added selectivity for a wide range of compounds and improved robustness.