Learn More
Waters Corp Column, 2-PIC, 130Å, 5 μm, 4.6 mm × 100 mm
Catalog No. 502390613 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns are designed for scaling SuperCritical Fluid (SFC) applications from analytical to preparative. The column chemistries, including 2-PIC, offer a wide range of selectivity and are available in various analytical and preparative formats.
  • Streamline purchasing with customized quotes.
  • Offers a wide range of selectivity when scaling SFC from analytical to preparative.
  • Available in 1.7 μm and 5 μm analytical and 5 μm preparative column formats.
Catalog No. 50-239-0613 Supplier Waters Corp Supplier No. 186008551
These columns are designed for scaling SuperCritical Fluid (SFC) applications from analytical to preparative. The column chemistries, including 2-PIC, offer a wide range of selectivity and are available in various analytical and preparative formats.
  • Streamline purchasing with customized quotes.
  • Offers a wide range of selectivity when scaling SFC from analytical to preparative.
  • Available in 1.7 μm and 5 μm analytical and 5 μm preparative column formats.