Learn More
Waters Corp Waters Corporation Torus Column, 2-PIC, 1.7 μm, 130 Å, 2.1 x 50 mm
Catalog No. NC2223615 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Torus 2-PIC Column is an analytical supercritical fluid chromatography (SFC) column, featuring spherical particles with a 1.7 μm particle size, 0.7 cc/g pore volume, 130 Å pore size, and 185 m²/g surface area. With dimensions of 2.1 x 50 mm, its 2-Picolylamine chemistry makes it ideal for general use with a wide range of acidic and basic compounds.
  • Provides improved peak shape, added retention, and novel selectivity compared to conventional 2-ethylpyridine (2-EP) columns.
  • Designed for general use in SFC separations for a wide range of acidic and basic compounds.
  • Demonstrates excellent peak shape characteristics for diverse acidic and basic compounds.
  • Designed for robustness, providing consistent chromatographic performance over the lifetime of the column.
  • Suitable for method transfer and method scale-up processes.
Catalog No. NC2223615 Supplier Waters Corp Supplier No. 186007596
The Torus 2-PIC Column is an analytical supercritical fluid chromatography (SFC) column, featuring spherical particles with a 1.7 μm particle size, 0.7 cc/g pore volume, 130 Å pore size, and 185 m²/g surface area. With dimensions of 2.1 x 50 mm, its 2-Picolylamine chemistry makes it ideal for general use with a wide range of acidic and basic compounds.
  • Provides improved peak shape, added retention, and novel selectivity compared to conventional 2-ethylpyridine (2-EP) columns.
  • Designed for general use in SFC separations for a wide range of acidic and basic compounds.
  • Demonstrates excellent peak shape characteristics for diverse acidic and basic compounds.
  • Designed for robustness, providing consistent chromatographic performance over the lifetime of the column.
  • Suitable for method transfer and method scale-up processes.