Learn More
Waters Corp Waters Corporation Torus 2-PIC OBD Prep Column, 5 μm, 130 Å, 10 × 250 mm
Supplier:  Waters Corp 186008584
Encompass_Preferred
The Torus 2-PIC OBD Prep Column, designed for SuperCritical Fluid (SFC) analytical to preparative scaling, offers wide selectivity and robust, dependable achiral separations with ACQUITY UPC2 System.
  • SFC analytical to preparative scaling.
  • Wide selectivity across formats.
  • Fast, dependable achiral separations (ACQUITY UPC2).
  • Advanced chemistries: excellent peak shape, improved selectivity.
  • High robustness; effective for acidic/basic compounds.
  • ISO-certified manufacturing, batch tested.
  • Performance Chromatogram and Batch Analysis Certificate included.
  • Key Specs: Separation: SFC; Substrate: Hybrid; Max pressure: 6000 psi (415 Bar); Temp: 60°C; Shape: Spherical; Surface area: 185.
Catalog No. 50-203-8672