Learn More
Waters Corp Torus 2-PIC OBD Prep Column, 130Å Pore, 5 μm, 50 × 150 mm
Catalog No. 502390608 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Torus 2-PIC OBD Prep Column is engineered to enhance the efficiency of the ACQUITY UPC2 system for swift and dependable achiral separations. This column simplifies the method development process, provides excellent peak shape, and reduces or eliminates the need for additives, while offering additional selectivity for various compounds and improving robustness.
  • Specifically designed to utilize the full power of the ACQUITY UPC2 system for fast and robust achiral separations.
  • Simplifies the method development process.
  • Provides excellent peak shape.
  • Eliminates or reduces the need for additives.
  • Offers additional selectivity for a broad range of compounds.
  • Ensures improved robustness.
Catalog No. 50-239-0608 Supplier Waters Corp Supplier No. 186008596
Add to Cart
Add to Cart
Request a Quote
The Torus 2-PIC OBD Prep Column is engineered to enhance the efficiency of the ACQUITY UPC2 system for swift and dependable achiral separations. This column simplifies the method development process, provides excellent peak shape, and reduces or eliminates the need for additives, while offering additional selectivity for various compounds and improving robustness.
  • Specifically designed to utilize the full power of the ACQUITY UPC2 system for fast and robust achiral separations.
  • Simplifies the method development process.
  • Provides excellent peak shape.
  • Eliminates or reduces the need for additives.
  • Offers additional selectivity for a broad range of compounds.
  • Ensures improved robustness.