Learn More
Waters Corp Torus OBD Preparative Column, 2-PIC, 5 μm, 130 Å, 30 mm X 75 mm
Catalog No. 502391093 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column enables the scaling of SuperCritical Fluid (SFC) from analytical to preparative applications. It is part of the Torus column chemistries, which offer a broad range of selectivity, including 2-PIC, DIOL, DEA, and 1-AA. These columns are available in both analytical (1.7 μm and 5 μm) and preparative (5 μm) formats.
  • Offers a wide range of selectivity through various column chemistries (2-PIC, DIOL, DEA, 1-AA).
  • Supports SFC scaling from analytical to preparative applications.
  • Available in different particle sizes and formats to suit various analytical and preparative needs.
  • Designed as an OBD (Optimum Bed Density) preparative column.
Catalog No. 50-239-1093 Supplier Waters Corp Supplier No. 186008590
Add to Cart
Add to Cart
Request a Quote
This column enables the scaling of SuperCritical Fluid (SFC) from analytical to preparative applications. It is part of the Torus column chemistries, which offer a broad range of selectivity, including 2-PIC, DIOL, DEA, and 1-AA. These columns are available in both analytical (1.7 μm and 5 μm) and preparative (5 μm) formats.
  • Offers a wide range of selectivity through various column chemistries (2-PIC, DIOL, DEA, 1-AA).
  • Supports SFC scaling from analytical to preparative applications.
  • Available in different particle sizes and formats to suit various analytical and preparative needs.
  • Designed as an OBD (Optimum Bed Density) preparative column.