Learn More
Waters Corp OBD Prep Column, 130Å, 5 μm, 30 mm, 250 mm
Catalog No. 502391428 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns allow for scaling SuperCritical Fluid Chromatography (SFC) from analytical to preparative applications.
  • Scale SFC from analytical to preparative.
  • Column chemistries offer a wide range of selectivity.
  • Available in various analytical and preparative column formats, including 1.7 μm and 5 μm particle sizes.
Catalog No. 50-239-1428 Supplier Waters Corp Supplier No. 186008593
Add to Cart
Add to Cart
Request a Quote
These columns allow for scaling SuperCritical Fluid Chromatography (SFC) from analytical to preparative applications.
  • Scale SFC from analytical to preparative.
  • Column chemistries offer a wide range of selectivity.
  • Available in various analytical and preparative column formats, including 1.7 μm and 5 μm particle sizes.