Learn More
Waters Corp Torus 2-PIC OBD Prep Column, 130Å, 5 μm, 50 × 50 mm
Catalog No. 502391607 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Torus columns facilitate the scaling of Supercritical Fluid Chromatography (SFC) from analytical to preparative applications. The Torus 2-PIC column is a primary choice for a broad range of applications involving acidic and basic compounds, offering improved peak shape, enhanced retention, and novel selectivity compared to conventional 2-ethylpyridine (2-EP) phases. These columns utilize state-of-the-art media manufacturing and industry-leading column technology.
  • Imparts a new level of robustness to laboratory-scale purification.
  • Allows for the scale-up of screening methods from 1.7 μm analytical columns.
  • Based on patent-pending two-stage functionalization of ethylene bridged hybrid (BEH) particles for broad selectivities and robust chromatographic performance.
  • Excellent peak shapes.
  • Highest efficiency and QC-ready robustness.
  • Utilizes OBD technology.
Catalog No. 50-239-1607 Supplier Waters Corp Supplier No. 186008594
Add to Cart
Add to Cart
Request a Quote
Torus columns facilitate the scaling of Supercritical Fluid Chromatography (SFC) from analytical to preparative applications. The Torus 2-PIC column is a primary choice for a broad range of applications involving acidic and basic compounds, offering improved peak shape, enhanced retention, and novel selectivity compared to conventional 2-ethylpyridine (2-EP) phases. These columns utilize state-of-the-art media manufacturing and industry-leading column technology.
  • Imparts a new level of robustness to laboratory-scale purification.
  • Allows for the scale-up of screening methods from 1.7 μm analytical columns.
  • Based on patent-pending two-stage functionalization of ethylene bridged hybrid (BEH) particles for broad selectivities and robust chromatographic performance.
  • Excellent peak shapes.
  • Highest efficiency and QC-ready robustness.
  • Utilizes OBD technology.