Learn More
Waters Corp Prep Guard Cartridge, 130Å, 5 μm, 19 mm X 10 mm
Catalog No. 502391016 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This guard cartridge is designed to extend column lifetime by protecting prep columns from particulates and contaminants in the mobile phase. It features a 130Å, 5μm sorbent bed bonded to a hybrid silica substrate, ensuring exceptional retention of impurities while preserving target analytes. Compatible with all SFC Systems, it supports diverse applications and integrates seamlessly into laboratory workflows.
  • Extends column lifetime
  • Protects prep columns from particulates and contaminants
  • 130Å, 5μm sorbent bed with hybrid silica substrate
  • Ensures retention of impurities while preserving target analytes
  • Compatible with all SFC systems
  • Supports diverse applications like lipid analysis, oligonucleotide analysis, and metabolomics
  • User-friendly installation
  • Recommended replacement every 100 to 200 injections
Catalog No. 50-239-1016 Supplier Waters Corp Supplier No. 186008741
This guard cartridge is designed to extend column lifetime by protecting prep columns from particulates and contaminants in the mobile phase. It features a 130Å, 5μm sorbent bed bonded to a hybrid silica substrate, ensuring exceptional retention of impurities while preserving target analytes. Compatible with all SFC Systems, it supports diverse applications and integrates seamlessly into laboratory workflows.
  • Extends column lifetime
  • Protects prep columns from particulates and contaminants
  • 130Å, 5μm sorbent bed with hybrid silica substrate
  • Ensures retention of impurities while preserving target analytes
  • Compatible with all SFC systems
  • Supports diverse applications like lipid analysis, oligonucleotide analysis, and metabolomics
  • User-friendly installation
  • Recommended replacement every 100 to 200 injections