Learn More
Waters Corp Torus 2-PIC OBD Prep Column, 130Å, 5 µm, 19 mm X 250 mm, 1/pk
Catalog No. NC1816930 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Scale SFC from analytical to preparative with Torus Columns. Torus column chemistries (Torus 2-PIC, Torus DIOL, Torus DEA and Torus 1-AA) offer a wide range of selectivity and are offered in 1.7 µm and 5 µm analytical and 5 µm preparative column formats.
Catalog No. NC1816930 Supplier Waters Corp Supplier No. 186008588
Add to Cart
Add to Cart
Request a Quote
Scale SFC from analytical to preparative with Torus Columns. Torus column chemistries (Torus 2-PIC, Torus DIOL, Torus DEA and Torus 1-AA) offer a wide range of selectivity and are offered in 1.7 µm and 5 µm analytical and 5 µm preparative column formats.