Learn More
Waters Corp ACQUITY UPC2 Torus DEA Column, 1.7 μm Particle Size, 2.1 × 75 mm, Diethylamine
Catalog No. 502391756 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPC2 Torus DEA Column is engineered to deliver superior peak shape for very strong bases, offering complementary selectivity to other phases. This column provides excellent chromatographic performance for strong basic compounds, even without the need for additives.
  • Offers selectivity that complements other phases.
  • Designed to provide superior peak shape for very strong bases, with or without additives.
  • Achieves excellent peak shape for strong basic compounds without additives, outperforming standard silica columns.
Catalog No. 50-239-1756 Supplier Waters Corp Supplier No. 186007615
The ACQUITY UPC2 Torus DEA Column is engineered to deliver superior peak shape for very strong bases, offering complementary selectivity to other phases. This column provides excellent chromatographic performance for strong basic compounds, even without the need for additives.
  • Offers selectivity that complements other phases.
  • Designed to provide superior peak shape for very strong bases, with or without additives.
  • Achieves excellent peak shape for strong basic compounds without additives, outperforming standard silica columns.