Learn More
Waters Corp Column, DEA, 130Å, 5 μm, 2.1 mm, 150 mm
Supplier:  Waters Corp 186008563
Encompass_Preferred
The Torus DEA Column is engineered for excellent peak shape and increased selectivity, facilitating quick and robust achiral separations. It utilizes novel 1.7 μm chemistry for convergence chromatography and revolutionary bonding technology for adaptable selectivity.
  • Engineered for excellent peak shape, reducing or avoiding the need for additives.
  • Provides increased selectivity for a wide range of chemicals.
  • Offers greater resilience.
  • Enables quick, robust achiral separations.
  • Utilizes the full spectrum of system capabilities.
  • Features novel and revolutionary 1.7 μm chemistry for convergence chromatography.
  • Streamlines the method development process.
  • Based on revolutionary bonding technology for an adaptable selectivity platform.
Catalog No. 50-239-1454