Learn More
Waters Corp Torus DEA OBD Preparative Column, 50 × 100 mm, 5 μm, 130 Å
Catalog No. 502390662 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Torus DEA OBD Preparative Column is designed for achiral SFC separations, offering excellent peak shapes and unique selectivity with a diethylamine ligand. It utilizes OBD Technology for high efficiency and robustness, and features a hydrophilic surface for controlled retention and minimized unwanted interactions.
  • Designed for achiral SFC separations.
  • Offers excellent peak shapes.
  • Provides unique selectivity with a diethylamine ligand.
  • Exhibits high efficiency and QC-ready robustness.
  • Utilizes OBD Technology.
  • Orthogonal to Torus 2-PIC phase.
  • Provides superior peak shape for very strong bases.
  • Based on patent-pending two-stage functionalization of ethylene bridged hybrid (BEH) particles.
  • Hydrophilic surface controls retention and minimizes unwanted surface interactions.
Catalog No. 50-239-0662 Supplier Waters Corp Supplier No. 186008629
Add to Cart
Add to Cart
Request a Quote
The Torus DEA OBD Preparative Column is designed for achiral SFC separations, offering excellent peak shapes and unique selectivity with a diethylamine ligand. It utilizes OBD Technology for high efficiency and robustness, and features a hydrophilic surface for controlled retention and minimized unwanted interactions.
  • Designed for achiral SFC separations.
  • Offers excellent peak shapes.
  • Provides unique selectivity with a diethylamine ligand.
  • Exhibits high efficiency and QC-ready robustness.
  • Utilizes OBD Technology.
  • Orthogonal to Torus 2-PIC phase.
  • Provides superior peak shape for very strong bases.
  • Based on patent-pending two-stage functionalization of ethylene bridged hybrid (BEH) particles.
  • Hydrophilic surface controls retention and minimizes unwanted surface interactions.