Learn More
Waters Corp Torus DEA OBD Prep Column, 130 Å, 5 μm, 19 mm × 100 mm
Catalog No. 502392668 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Torus DEA OBD Prep Column is designed for preparative chromatography applications. It features Diethylamine chemistry and a spherical particle shape for efficient separation.
  • Designed to be orthogonal to the Torus 2-PIC phase.
  • Provides superior peak shape for very strong bases.
  • Offers complementary selectivity to the 2-PIC stationary phase.
Catalog No. 50-239-2668 Supplier Waters Corp Supplier No. 186008620
The Torus DEA OBD Prep Column is designed for preparative chromatography applications. It features Diethylamine chemistry and a spherical particle shape for efficient separation.
  • Designed to be orthogonal to the Torus 2-PIC phase.
  • Provides superior peak shape for very strong bases.
  • Offers complementary selectivity to the 2-PIC stationary phase.