Learn More
Waters Corp Torus DEA OBD Prep Column, 130Å, 5 μm, 19 × 50 mm
Catalog No. 502392107 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Torus DEA OBD Prep Column is designed for preparative chromatography applications. It features a unique chemistry tailored for specific separations, providing high efficiency and reproducibility.
  • 130Å pore size allows for effective separation of various compounds.
  • 5 μm particle size ensures high resolution and efficient separations.
  • Column dimensions of 19 mm × 50 mm are suitable for preparative scale work.
  • Optimized for high-throughput purification and robust performance.
Catalog No. 50-239-2107 Supplier Waters Corp Supplier No. 186008619
Add to Cart
Add to Cart
This Torus DEA OBD Prep Column is designed for preparative chromatography applications. It features a unique chemistry tailored for specific separations, providing high efficiency and reproducibility.
  • 130Å pore size allows for effective separation of various compounds.
  • 5 μm particle size ensures high resolution and efficient separations.
  • Column dimensions of 19 mm × 50 mm are suitable for preparative scale work.
  • Optimized for high-throughput purification and robust performance.